missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trefoil factor 3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23277-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Beschreibung
Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing. Trefoil peptides contain three intrachain disulfide bonds, forming the trefoil motif, or P-domain and are clustered in human chromosome 21q22.3. Trefoil Factor 3 (TFF3) is one of three trefoil peptides secreted by epithelial cells that line mucus membranes in the large and small intestines and lower respiratory tract. TFF3 is mainly active as a disulfide-linked homodimer and may have a role in promoting cell migration in healing processes. It is involved in the maintece and repair of the intestinal mucosa.Spezifikation
Trefoil factor 3 | |
Polyclonal | |
Unconjugated | |
TFF3 | |
HITF, hP1.B, Intestinal trefoil factor, ITF, Polypeptide P1.B, TFF3, TFI, Trefoil factor 3, trefoil factor 3 (intestinal) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7033 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q07654 | |
TFF3 | |
Trefoil factor 3 Fusion Protein Ag9543 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |