Learn More
TRIM21 Mouse anti-Human, Unconjugated, Clone: 1E8B9, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67136-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16829595
  • 121.38€ / 20µL
Expédition estimée: 03-10-2022
Description
The TRIM21 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM21. This antibody recognizes human, mouse, rat antigen. The TRIM21 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
TRIM21
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Rat, Human
Immunohistochemistry, Immunoprecipitation, Western Blot
1E8B9
Unconjugated
20 μL
6737
IgG1