missing translation for 'onlineSavingsMsg'
Learn More

TRIM5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 11476-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16828583

  • 325.38€ / 150µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene is a member of the tripartite motif family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein forms homo-oligomers via the coiled-coil region and localizes to cytoplasmic bodies. It appears to function as a E3 ubiquitin-ligase and ubiquitinates itself to regulate its subcellular localization. It may play a role in retroviral restriction. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Spécification

Spécification

TRIM5
Polyclonal
Unconjugated
TRIM5
RING finger protein 88, RNF88, TRIM5, TRIM5alpha, tripartite motif containing 5
Rabbit
Antigen Affinity Chromatography
RUO
667823, 85363
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9C035
TRIM5
TRIM5 Fusion Protein Ag1973
150 μL
Primary
Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions