missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM67 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24369-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The specific function of TRIM67 is not yet known.Spécification
TRIM67 | |
Polyclonal | |
Unconjugated | |
TRIM67 | |
TNL, TRIM67, TRIM9 like protein, tripartite motif containing 67 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
330863, 440730 | |
-20°C | |
Liquid |
Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q505D9, Q6ZTA4 | |
Trim67 | |
TRIM67 Fusion Protein Ag21632 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |