missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13722-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the SWI2/SNF2 family of proteins, which play a critical role in altering protein-DNA interactions. The encoded protein has been shown to have dsDNA-dependent ATPase activity and RNA polymerase II termination activity. This protein interacts with cell division cycle 5-like, associates with human splicing complexes, and plays a role in pre-mRNA splicing.Spécification
TTF2 | |
Polyclonal | |
Unconjugated | |
TTF2 | |
F2, HuF2, Lodestar homolog, Transcription release factor 2, TTF2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
295324, 74044, 8458 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5NC05, Q9UNY4 | |
Ttf2 | |
TTF2 Fusion Protein Ag4671 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |