missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTL Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2E5F8, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66076-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Catalyzes the post-translational addition of a tyrosine to the C-terminal end of detyrosinated alpha-tubulin.Spécification
TTL | |
Monoclonal | |
0.76 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P38160, P38585, Q8NG68, Q9QXJ0 | |
Ttl | |
TTL Fusion Protein Ag4708 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
2E5F8 | |
Unconjugated | |
TTL | |
TTL, Tubulin tyrosine ligase, tubulin tyrosine ligase | |
Mouse | |
Protein A | |
RUO | |
150465, 171572, 445530, 69737 | |
-20°C | |
Liquid |