missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTPA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27081-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a soluble protein that binds alpha-trocopherol, a form of vitamin E, with high selectivity and affinity. This protein plays an important role in regulating vitamin E levels in the body by transporting vitamin E between membrane vesicles and facilitating the secretion of vitamin E from hepatocytes to circulating lipoproteins. Mutations in this gene cause hereditary vitamin E deficiency (ataxia with vitamin E deficiency, AVED) and retinitis pigmentosa.Spécification
TTPA | |
Polyclonal | |
Unconjugated | |
TTPA | |
Alpha TTP, alphaTTP, ATTP, AVED, TPP1, TTP1, TTPA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25571, 50500, 7274 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P41034, P49638, Q8BWP5 | |
Ttpa | |
TTPA Fusion Protein Ag24177 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |