missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2S Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 18219-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the ubiquitin-conjugating enzyme family. The encoded protein is able to form a thiol ester linkage with ubiquitin in a ubiquitin activating enzyme-dependent manner, a characteristic property of ubiquitin carrier proteins.Spécification
UBE2S | |
Polyclonal | |
Unconjugated | |
UBE2S | |
E2 EPF, E2EPF, EPF5, UBE2S, Ubiquitin carrier protein S, Ubiquitin protein ligase S | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27338, 292588, 77891 | |
-20°C | |
Liquid |
Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B5DFI8, Q16763, Q921J4 | |
UBE2S | |
UBE2S Fusion Protein Ag12978 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |