missing translation for 'onlineSavingsMsg'
Learn More

VAMP7/TI-VAMP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 22268-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16811815

  • 140.00€ / 20µL
Expédition estimée: 03-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The antibody is specific to VAMP7.

This gene encodes a transmembrane protein that is a member of the soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) family. The encoded protein localizes to late endosomes and lysosomes and is involved in the fusion of transport vesicles to their target membranes. Alternate splicing results in multiple transcript variants.
Spécification

Spécification

VAMP7/TI-VAMP
Polyclonal
Unconjugated
VAMP7
SYBL1, Synaptobrevin like protein 1, Tetanus insensitive VAMP, TI VAMP, TI-VAMP, VAMP 7, VAMP7
Rabbit
Antigen Affinity Chromatography
RUO
20955, 6845, 85491
-20°C
Liquid
Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
0.6 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P51809, P70280, Q9JHW5
Vamp7
Peptide
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions