Learn More
VEGFA Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26381-1-AP-20UL
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
The VEGFA antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human VEGFA. This antibody recognizes human antigen. The VEGFA antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Spécification
VEGFA | |
Polyclonal | |
Unconjugated | |
20 μL | |
7422 | |
IgG |
Western Blot, Immunohistochemistry, Flow Cytometry, Co-immunoprecipitation (Co-IP), ELISA | |
233 μg/mL | |
Rabbit | |
Primary | |
Human |