missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vpr Rabbit anti-Virus, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 51143-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody recognize the VPR gene of Human immunodeficiency virus 1.
Spécification
Vpr | |
Polyclonal | |
Unconjugated | |
Vpr | |
Vpr Fusion Protein Ag0718 | |
20 μL | |
Primary | |
-20°C | |
Liquid |
ELISA, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |