Learn More
VPS35 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 10236-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16836493
  • 121.38€ / 20µL
Expédition estimée: 30-09-2022
Description
The VPS35 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VPS35. This antibody recognizes human, mouse, rat antigen. The VPS35 antibody has been validated for the following applications: IF, IP, WB, ELISA analysis.
Spécification
Spécification
VPS35
Polyclonal
Unconjugated
20 μL
55737
IgG
CoIP, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
260 μg/mL
Rabbit
Primary
Mouse, Rat, Human