missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VTA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15786-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
C6ORF55 encodes a protein involved in trafficking of the multivesicular body, an endosomal compartment involved in sorting membrane proteins for degradation in lysosomes.Spécification
VTA1 | |
Polyclonal | |
Unconjugated | |
VTA1 | |
C6orf55, DRG 1, DRG1, HSPC228, LIP5, LYST interacting protein 5, My012, SBP1, SKD1 binding protein 1, VTA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51534 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NP79 | |
VTA1 | |
VTA1 Fusion Protein Ag8485 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |