missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WHSC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22722-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a protein that contains four domains present in other developmental proteins: a PWWP domain, an HMG box, a SET domain, and a PHD-type zinc finger. It is expressed ubiquitously in early development. Wolf-Hirschhorn syndrome (WHS) is a malformation syndrome associated with a hemizygous deletion of the distal short arm of chromosome 4. This gene maps to the 165 kb WHS critical region and has also been involved in the chromosomal translocation t(4;14)(p16.3;q32.3) in multiple myelomas. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms. Some transcript variants are nonsense-mediated mRNA (NMD) decay candidates, hence not represented as reference sequences.Spécification
WHSC1 | |
Polyclonal | |
Unconjugated | |
NSD2 | |
KIAA1090, MMSET, NSD2, Protein trithorax 5, REIIBP, TRX5, WHS, WHSC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7468 | |
-20°C | |
Liquid |
Immunoprecipitation | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O96028 | |
NSD2 | |
WHSC1 Fusion Protein Ag18611 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |