Learn More
WISP3 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21259-1-AP-20UL
Informations supplémentaires : Poids : 0.00020kg
Description
The WISP3 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human WISP3. This antibody recognizes human antigen. The WISP3 antibody has been validated for the following applications: WB,ELISA analysis.
Spécification
WISP3 | |
Polyclonal | |
Unconjugated | |
20 μL | |
8838 | |
Liquid |
Western Blot, ELISA | |
350 μg/mL | |
Rabbit | |
Primary | |
Human |