Learn More
WISP3 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 21259-1-AP-20UL
Informations supplémentaires : Poids : 0.00020kg
Code produit. 17271366
  • 140.00€ / 20µL
Expédition estimée: 24-05-2024
Description
The WISP3 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human WISP3. This antibody recognizes human antigen. The WISP3 antibody has been validated for the following applications: WB,ELISA analysis.
Spécification
Spécification
WISP3
Polyclonal
Unconjugated
20 μL
8838
Liquid
Western Blot, ELISA
350 μg/mL