missing translation for 'onlineSavingsMsg'
Learn More

ZC3H8 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 26152-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16834685

  • 325.38€ / 150µL
Expédition estimée: 10-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Zinc-finger CCCH domain-containing protein 8 (ZC3H8) acts as a transcriptional repressor of the GATA3 promoter. It is a sequence-specific DNA-binding factor that binds to the 5′-AGGTCTC-3′ sequence within the negative cis-acting element intronic regulatory region (IRR) of the GATA3 gene. ZC3H8 induces thymocyte apoptosis when overexpressed, which may indicate a role in regulation of thymocyte homeostasis.
Spécification

Spécification

ZC3H8
Polyclonal
Unconjugated
ZC3H8
Fliz1, ZC3H8, ZC3HDC8
Rabbit
Antigen Affinity Chromatography
RUO
84524
-20°C
Liquid
Western Blot
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8N5P1
ZC3H8
ZC3H8 Fusion Protein Ag23514
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions