missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF134 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25515-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppeltype DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc-finger protein 134 (ZNF134) is a 348 amino acid member of the Krueppel C2H2-type zinc-finger protein family. ZNF134 localizes to the nucleus and contains nine C2H2-type zinc fingers through which it is thought to be involved in DNA-binding and transcriptional regulation.Spécification
ZNF134 | |
Polyclonal | |
Unconjugated | |
ZNF134 | |
pHZ 15, zinc finger protein 134, ZNF134 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7693 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P52741 | |
ZNF134 | |
ZNF134 Fusion Protein Ag22290 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |