missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF192 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21656-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes one of four subunits of the splicing factor 3B. The protein encoded by this gene cross-links to a region in the pre-mRNA immediately upstream of the branchpoint sequence in pre-mRNA in the prespliceosomal complex A. It also may be involved in the assembly of the B, C and E spliceosomal complexes. In addition to RNA-binding activity, this protein interacts directly and highly specifically with subunit 2 of the splicing factor 3B. This protein contains two N-terminal RNA-recognition motifs (RRMs), consistent with the observation that it binds directly to pre-mRNA.Spécification
ZNF192 | |
Polyclonal | |
Unconjugated | |
ZKSCAN8 | |
LD5 1, zinc finger protein 192, ZKSCAN8, ZNF192 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7745, 93681 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15776 | |
Zkscan8 | |
ZNF192 Fusion Protein Ag16381 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |