missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-c-Cbl (Tyr774) Recombinant Rabbit Monoclonal Antibody (CblY774-R3B8), APC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA537104
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Cbl proteins are a family of ubiquitin protein ligases (E3s) that negatively regulate signaling by targeting activated tyrosine kinases for degradation. Cbl- c is the most recently cloned member of the Cbl proteins and is expressed only in epithelial cells (the other Cbl proteins are ubiquitously expressed). Cbl-c, like the other mammalian Cbl proteins, can ubiquitinate the activated EGFR and target it for degradation. Through interactions with proteins containing SRC homology-2 (SH2) and SH3 domains, CBL proteins modulate downstream cell signaling.
Tekniske data
Phospho-c-Cbl (Tyr774) | |
Recombinant Monoclonal | |
APC | |
Cbl | |
4732447J05Rik; Cas-Br-M (murine) ecotropic retroviral transforming sequence; Casitas B-lineage lymphoma; casitas B-lineage lymphoma proto-oncogene; CBL; Cbl proto-oncogene; Cbl proto-oncogene, E3 ubiquitin protein ligase; CBL2; Cbl-2; CBL-3; CBLC; c-Cbl; E3 ubiquitin-protein ligase CBL; FRA11B; fragile site, folic acid type, rare, fra(11)(q23.3); NSLL; oncogene CBL2; proto-oncogene c-Cbl; RGD1561386; RING finger protein 55; RING-type E3 ubiquitin transferase CBL; RNF55; RNF57; signal transduction protein CBL | |
Rabbit | |
Protein A/G | |
RUO | |
12402, 867 | |
4°C | |
Liquid |
Flow Cytometry | |
CblY774-R3B8 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
P22681, P22682 | |
Cbl | |
A synthetic phospho-peptide corresponding to residues surrounding Tyr774 of human phospho c-Cbl. | |
100 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |
Produktforslag
Clients qui ont consulté cet article ont également consulté
Viewing 1-4 of
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Invitrogen™ Phospho-c-Cbl (Tyr774) Recombinant Rabbit Monoclonal Antibody (CblY774-R3B8), APC, Invitrogen™ > 100 Tests; APC
Ser du en mulighed for forbedring?Del en indholdskorrektion